2,7-Dibromo-9H-thioxanthen-9-one 98%

Supplier: Ambeed
Warning

A1442327-1G A1442327-250MG A1442327-100MG
76661-060EA 72.15 USD
76661-060 76661-062 77622-100
2,7-Dibromo-9H-thioxanthen-9-one 98%
2,7-Dibromo-9H-thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₃H₆Br₂OS
MW: 370.0591 g/mol
MDL Number: MFCD32880073
CAS Number: 40102-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR