2-Bromo-1,1-diethoxypropane 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopriopionaldehydediethylacetal

A1442096-1G A1442096-25G A1442096-5G
76661-044EA 26.53 USD
76661-044 76661-046 76661-048
2-Bromo-1,1-diethoxypropane 95%
2-Bromo-1,1-diethoxypropane

Precaución: for Research Use Only

Formula: (H₃CCH₂O)₂HCBrCHCH₃
MW: 211.1 g/mol
MDL Number: MFCD00152299
CAS Number: 3400-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR