4-Bromo-5-chloronaphthalen-2-ol 97%

Supplier: Ambeed
Warning

A1442088-1G A1442088-250MG A1442088-5G
76661-034EA 182 USD
76661-034 77874-837 77874-838
4-Bromo-5-chloronaphthalen-2-ol 97%
4-Bromo-5-chloronaphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₆BrClO
CAS Number: 2454397-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR