Bromophenol blue sodium salt indicator

Supplier: Ambeed
Warning

A1441854-25G A1441854-5G A1441854-100G
76661-020EA 27 USD
76661-020 76661-022 77597-196
Bromophenol blue sodium salt indicator
Bromophenol blue sodium salt

Precaución: for Research Use Only

Formula: C₁₉H₉Br₄NaO₅S
MDL Number: MFCD00013793
CAS Number: 34725-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR