4-(4-(1-((4-Formylbenzoyl)oxy)ethyl)-2-methoxy-5-nitrophenoxy)butanoic acid 97%

Supplier: Ambeed
Warning

A1441794-100MG A1441794-250MG A1441794-50MG
76661-014EA 224.92 USD
76661-014 76661-016 76661-018
4-(4-(1-((4-Formylbenzoyl)oxy)ethyl)-2-methoxy-5-nitrophenoxy)butanoic acid 97%
4-(4-(1-((4-Formylbenzoyl)oxy)ethyl)-2-methoxy-5-nitrophenoxy)butanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₉
MW: 431.3928 g/mol
MDL Number: MFCD34186933
CAS Number: 2703752-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR