Diethyl 2-(6-hydroxyhexyl)malonate 90%

Supplier: Ambeed
Warning

A1441614-250MG A1441614-100MG
76661-006EA 152.77 USD
76661-006 77597-192
Diethyl 2-(6-hydroxyhexyl)malonate 90%
Diethyl 2-(6-hydroxyhexyl)malonate

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₅
MW: 260.3267 g/mol
CAS Number: 108051-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR