3-(4-Iodo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 95%
Supplier: Ambeed
|
Warning
|
A1441561-250MG
A1441561-5G
A1441561-1G
A1441561-100MG
76661-002EA
59
USD
76661-002
76661-004
77592-706
77622-086
3-(4-Iodo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 95%
3-(4-Iodo-1-oxoisoindolin-2-yl)piperidine-2,6-dione
Precaución: for Research Use Only
Formula:
C₁₃H₁₁IN₂O₃ |
MDL Number:
MFCD32862119 CAS Number: 2207541-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...