4-Bromo-7-fluorobenzo[d]thiazol-2-amine 97%
Supplier: Ambeed
|
Warning
|
A1441092-100MG
A1441092-10G
A1441092-1G
A1441092-250MG
A1441092-5G
76660-980EA
63.66
USD
76660-980
76660-982
76660-984
76660-986
76660-988
4-Bromo-7-fluorobenzo[d]thiazol-2-amine 97%
4-Bromo-7-fluorobenzo[d]thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₇H₄BrFN₂S MW: 247.0875 g/mol |
MDL Number:
MFCD09475480 CAS Number: 942473-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...