(T-4)-[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]zinc 95%

Supplier: Ambeed
Danger

A1440858-250MG A1440858-100MG A1440858-1G A1440858-5G
76660-964EA 47.75 USD
76660-964 77622-078 77622-080 77622-082
(T-4)-[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]zinc 95%
(T-4)-[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]zinc

Precaución: for Research Use Only

Formula: C₃₆H₅₂N₂O₂Zn
MW: 610.2193 g/mol
CAS Number: 351498-19-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR