Nonacosan-15-ylbenzene 97%

Supplier: Ambeed
Warning

A1440826-100MG A1440826-250MG
76660-960EA 130.5 USD
76660-960 76660-962
Nonacosan-15-ylbenzene 97%
Nonacosan-15-ylbenzene

Precaución: for Research Use Only

Formula: C₃₅H₆₄
MW: 484.8827 g/mol
CAS Number: 56247-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR