(S)-3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione benzenesulfonate 97% ,99.99% ee

Supplier: Ambeed
Warning

A1440751-1G A1440751-250MG A1440751-100MG
76660-956EA 1039.69 USD
76660-956 76660-958 77622-076
(S)-3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione benzenesulfonate 97% ,99.99% ee
(S)-3-(1-Oxo-5-(piperazin-1-yl)isoindolin-2-yl)piperidine-2,6-dione benzenesulfonate

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₄O₆S
MW: 486.5407 g/mol
CAS Number: 2229714-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR