tert-Butyl (4-methylpyrrolidin-3-yl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A1440697-100MG A1440697-1G A1440697-250MG
76660-950EA 42 USD
76660-950 76660-952 76660-954
tert-Butyl (4-methylpyrrolidin-3-yl)carbamate hydrochloride 97%
tert-Butyl (4-methylpyrrolidin-3-yl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₂
MW: 236.7389 g/mol
MDL Number: MFCD32015928
CAS Number: 2682112-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR