6-(2-Methyl-5-nitrophenyl)-2-(methylthio)pyrido[3,4-d]pyrimidin-8(7H)-one 97%

Supplier: Ambeed
Warning

A1440348-1G A1440348-5G
76660-944EA 336.31 USD
76660-944 76660-946
6-(2-Methyl-5-nitrophenyl)-2-(methylthio)pyrido[3,4-d]pyrimidin-8(7H)-one 97%
6-(2-Methyl-5-nitrophenyl)-2-(methylthio)pyrido[3,4-d]pyrimidin-8(7H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₄O₃S
MW: 328.3458 g/mol
MDL Number: MFCD31977991
CAS Number: 2470433-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR