7,9-Bis(2,6-diisopropylphenyl)-7H-acenaphtho[1,2-d]imidazol-9-ium chloride 95%

Supplier: Ambeed
Warning

A1439975-100MG A1439975-1G A1439975-250MG A1439975-5G
76660-922EA 45.62 USD
76660-922 76660-924 76660-926 76660-928
7,9-Bis(2,6-diisopropylphenyl)-7H-acenaphtho[1,2-d]imidazol-9-ium chloride 95%
7,9-Bis(2,6-diisopropylphenyl)-7H-acenaphtho[1,2-d]imidazol-9-ium chloride

Precaución: for Research Use Only

Formula: C₃₇H₄₁ClN₂
MW: 549.1879 g/mol
CAS Number: 1246183-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR