2-(((Dodecylthio)carbonothioyl)thio)propanoic acid 97%

Supplier: Ambeed
Warning

A1439707-1G A1439707-250MG A1439707-25G A1439707-5G
76660-900EA 106.09 USD
76660-900 76660-902 76660-904 76660-906
2-(((Dodecylthio)carbonothioyl)thio)propanoic acid 97%
2-(((Dodecylthio)carbonothioyl)thio)propanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₀O₂S₃
MW: 350.6032 g/mol
MDL Number: MFCD22200510
CAS Number: 558484-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR