1-Methylcyclohexyl methacrylate 97%

Supplier: Ambeed
Warning

A1439615-1G A1439615-25G A1439615-5G
76660-896EA 17 USD
76660-896 76660-898 77874-807
1-Methylcyclohexyl methacrylate 97%
1-Methylcyclohexyl methacrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₂
MW: 182.2594 g/mol
CAS Number: 76392-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR