4-(Vinylsulfonyl)aniline 97+% stabilized
Supplier: Ambeed
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A1439532-100MG
A1439532-1G
A1439532-250MG
76660-888EA
185.66
USD
76660-888
76660-890
76660-892
4-(Vinylsulfonyl)aniline 97+% stabilized
4-(Vinylsulfonyl)aniline
Precaución: for Research Use Only
Formula:
C₈H₉NO₂S MW: 183.2276 g/mol |
MDL Number:
MFCD00460751 CAS Number: 25781-90-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...