(2,2'-Bipyridine)bis(1,10-phenanthroline)ruthenium(2+) bis(hexafluorophosphate) 97%
Supplier: Ambeed
|
Warning
|
A1439421-100MG
A1439421-250MG
A1439421-1G
76660-878EA
21
USD
76660-878
76660-880
77874-806
(2,2'-Bipyridine)bis(1,10-phenanthroline)ruthenium(2+) bis(hexafluorophosphate) 97%
(2,2'-Bipyridine)bis(1,10-phenanthroline)ruthenium(2+) bis(hexafluorophosphate)
Precaución: for Research Use Only
Formula:
C₃₄H₂₄F₆N₆PRu MW: 762.6288 g/mol |
CAS Number:
60828-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...