1,6-Dimethyl-1H-pyrazolo[4,3-d]pyrimidine-5,7(4H,6H)-dione 97%

Supplier: Ambeed
Warning

A1439417-100MG A1439417-1G A1439417-250MG
76660-872EA 178 USD
76660-872 76660-874 76660-876
1,6-Dimethyl-1H-pyrazolo[4,3-d]pyrimidine-5,7(4H,6H)-dione 97%
1,6-Dimethyl-1H-pyrazolo[4,3-d]pyrimidine-5,7(4H,6H)-dione

Precaución: for Research Use Only

Formula: C₇H₈N₄O₂
CAS Number: 56536-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR