6-Methoxypyrido[3,4-d]pyrimidin-4(1H)-one 97%

Supplier: Ambeed
Warning

A1439415-100MG A1439415-250MG
76660-868EA 188.85 USD
76660-868 76660-870
6-Methoxypyrido[3,4-d]pyrimidin-4(1H)-one 97%
6-Methoxypyrido[3,4-d]pyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.1600 g/mol
MDL Number: MFCD24597539
CAS Number: 183741-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR