N-Mesityl-1-methylpyrrolidine-2-carboxamide 97%

Supplier: Ambeed
Warning

A1439403-100MG A1439403-250MG
76660-854EA 122.01 USD
76660-854 76660-856
N-Mesityl-1-methylpyrrolidine-2-carboxamide 97%
N-Mesityl-1-methylpyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O
MW: 246.3480 g/mol
CAS Number: 30103-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR