1-((6-Chloropyridin-3-yl)methyl)imidazolidin-2-one 97%

Supplier: Ambeed
Warning

A1439327-100MG A1439327-1G A1439327-250MG
76660-844EA 96.55 USD
76660-844 76660-846 76660-848
1-((6-Chloropyridin-3-yl)methyl)imidazolidin-2-one 97%
1-((6-Chloropyridin-3-yl)methyl)imidazolidin-2-one

Precaución: for Research Use Only

Formula: C₉H₁₀ClN₃O
MDL Number: MFCD02661472
CAS Number: 120868-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR