4'-Fluoro-biphenyl-3-carbaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 4'-Fluoro-[1,1'-biphenyl]-3-carbaldehyde, 3-(4-Fluorophenyl)benzaldehyde

A1439143-100MG A1439143-1G A1439143-250MG A1439143-5G
76660-836EA 30.77 USD
76660-836 76660-838 76660-840 76660-842
4'-Fluoro-biphenyl-3-carbaldehyde 98%
4'-Fluoro-biphenyl-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.21 g/mol
MDL Number: MFCD01631878
CAS Number: 164334-74-1
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR