Methyl 3-(1,3,2-dioxaborinan-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A1438748-1G A1438748-5G
76660-832EA 37.14 USD
76660-832 76660-834
Methyl 3-(1,3,2-dioxaborinan-2-yl)benzoate 98%
Methyl 3-(1,3,2-dioxaborinan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₄
MW: 220.0295 g/mol
CAS Number: 2222401-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR