3-(1,3,2-Dioxaborinan-2-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A1438726-100MG A1438726-1G A1438726-250MG
76660-826EA 26.53 USD
76660-826 76660-828 76660-830
3-(1,3,2-Dioxaborinan-2-yl)benzoic acid 98%
3-(1,3,2-Dioxaborinan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₄
MW: 206.0029 g/mol
MDL Number: MFCD09839112
CAS Number: 2070921-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR