(11bS)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide 98%
Supplier: Ambeed
|
Warning
|
A1438407-100MG
A1438407-1G
A1438407-250MG
A1438407-5G
76660-818EA
127
USD
76660-818
76660-820
76660-822
76660-824
(11bS)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide 98%
(11bS)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide
Precaución: for Research Use Only
Formula:
C₆₈H₈₉O₄P |
CAS Number:
2757287-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...