4-Bromo-1-methyl-1H-imidazole-5-carbonitrile 97%

Supplier: Ambeed
Danger

A1438387-100MG A1438387-1G A1438387-250MG
76660-812EA 227.04 USD
76660-812 76660-814 76660-816
4-Bromo-1-methyl-1H-imidazole-5-carbonitrile 97%
4-Bromo-1-methyl-1H-imidazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄BrN₃
MW: 186.0094 g/mol
MDL Number: MFCD28642733
CAS Number: 54711-55-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR