(S)-tert-Butyl (1-hydroxy-3-methylbutan-2-yl)(methyl)carbamate 97%

Supplier: Ambeed
Warning

A1438385-1G A1438385-250MG
76660-810EA 195.21 USD
76660-810 77592-702
(S)-tert-Butyl (1-hydroxy-3-methylbutan-2-yl)(methyl)carbamate 97%
(S)-tert-Butyl (1-hydroxy-3-methylbutan-2-yl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₃NO₃
MW: 217.3052 g/mol
MDL Number: MFCD30069402
CAS Number: 251320-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR