Rel-(4aR,7aR)-octahydrothiopyrano[2,3-c]pyrrole 1,1-dioxide hydrochloride 98%

Supplier: Ambeed
Warning

A1438348-100MG A1438348-50MG
76660-804EA 137 USD
76660-804 76660-806
Rel-(4aR,7aR)-octahydrothiopyrano[2,3-c]pyrrole 1,1-dioxide hydrochloride 98%
Rel-(4aR,7aR)-octahydrothiopyrano[2,3-c]pyrrole 1,1-dioxide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂S
MDL Number: MFCD31560888
CAS Number: 2137857-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR