1-(trans-4-Aminocyclohexyl)ethanone hydrochloride 98%

Supplier: Ambeed
Warning

A1437657-100MG A1437657-250MG
76660-780EA 20 USD
76660-780 76660-782
1-(trans-4-Aminocyclohexyl)ethanone hydrochloride 98%
1-(trans-4-Aminocyclohexyl)ethanone hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO
MW: 177.6700 g/mol
MDL Number: MFCD31628951
CAS Number: 879876-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR