2-([1,1'-Biphenyl]-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1429478-1G A1429478-250MG A1429478-5G
76660-752EA 19.1 USD
76660-752 76660-754 76660-756
2-([1,1'-Biphenyl]-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane 98%
2-([1,1'-Biphenyl]-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₇H₁₉BO₂
MW: 266.1426 g/mol
CAS Number: 5123-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR