(4-(Benzo[d]oxazol-2-yl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1426791-1G A1426791-5G
76660-748EA 19 USD
76660-748 76660-750
(4-(Benzo[d]oxazol-2-yl)phenyl)boronic acid 98%
(4-(Benzo[d]oxazol-2-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BNO₃
MW: 239.0344 g/mol
MDL Number: MFCD32670117
CAS Number: 1065657-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR