1,2-Bis(4-bromophenyl)-1,2-diphenylethene 98%

Supplier: Ambeed
Warning

A1426724-1G A1426724-250MG A1426724-5G A1426724-10G
76660-742EA 34 USD
76660-742 76660-744 76660-746 77622-058
1,2-Bis(4-bromophenyl)-1,2-diphenylethene 98%
1,2-Bis(4-bromophenyl)-1,2-diphenylethene

Precaución: for Research Use Only

Formula: C₂₆H₁₈Br₂
MDL Number: MFCD01024668
CAS Number: 184239-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR