Methyl 3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%
Supplier: Ambeed
|
Warning
|
A1423837-1G
A1423837-250MG
A1423837-5G
76660-730EA
76.39
USD
76660-730
76660-732
76660-734
Methyl 3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%
Methyl 3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BO₄ MW: 248.0827 g/mol |
MDL Number:
MFCD32670192 CAS Number: 585524-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...