Methyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A1423819-100MG A1423819-1G A1423819-250MG A1423819-25G A1423819-5G
76660-704EA 8.49 USD
76660-704 76660-706 76660-708 76660-710 76660-712
Methyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%
Methyl 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₄
MW: 248.0827 g/mol
CAS Number: 905966-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR