1-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)ethan-1-one 98%

Supplier: Ambeed
Warning

A1420806-1G A1420806-250MG
76660-696EA 53.05 USD
76660-696 76660-698
1-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)ethan-1-one 98%
1-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₃
MW: 232.0833 g/mol
CAS Number: 460088-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR