Methyl 6-amino-2-chloro-5-methoxypyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A1417852-1G A1417852-250MG A1417852-5G
76660-678EA 66.84 USD
76660-678 76660-680 76660-682
Methyl 6-amino-2-chloro-5-methoxypyrimidine-4-carboxylate 98%
Methyl 6-amino-2-chloro-5-methoxypyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈ClN₃O₃
MW: 217.6097 g/mol
MDL Number: MFCD16652305
CAS Number: 1126320-49-7
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR