3-(1,3,2-Dioxaborinan-2-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

A1417378-1G A1417378-250MG A1417378-5G
76660-662EA 26.53 USD
76660-662 76660-664 76660-666
3-(1,3,2-Dioxaborinan-2-yl)benzaldehyde 98%
3-(1,3,2-Dioxaborinan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₃
MW: 190.0035 g/mol
CAS Number: 478930-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR