4-(1,3,2-Dioxaborinan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A1417376-100MG A1417376-1G
76660-660EA 20 USD
76660-660 77622-056
4-(1,3,2-Dioxaborinan-2-yl)benzonitrile 98%
4-(1,3,2-Dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
CAS Number: 152846-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR