3-(1,3,2-Dioxaborinan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A1417375-250MG A1417375-1G
76660-658EA 31 USD
76660-658 77622-054
3-(1,3,2-Dioxaborinan-2-yl)benzonitrile 98%
3-(1,3,2-Dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
CAS Number: 684648-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR