4-Chloro-N-methylquinazolin-2-amine 98%

Supplier: Ambeed
Warning

A1417127-100MG A1417127-1G A1417127-250MG
76660-652EA 74.27 USD
76660-652 76660-654 76660-656
4-Chloro-N-methylquinazolin-2-amine 98%
4-Chloro-N-methylquinazolin-2-amine

Precaución: for Research Use Only

Formula: C₉H₈ClN₃
MW: 193.6300 g/mol
MDL Number: MFCD29999981
CAS Number: 2090582-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR