4-((tert-Butyldimethylsilyl)oxy)butan-1-amine 98%

Supplier: Ambeed
Warning

A1415600-100MG A1415600-250MG A1415600-1G A1415600-5G
76660-646EA 57.29 USD
76660-646 76660-648 77566-848 77622-052
4-((tert-Butyldimethylsilyl)oxy)butan-1-amine 98%
4-((tert-Butyldimethylsilyl)oxy)butan-1-amine

Precaución: For research use only.

Formula: C₁₀H₂₅NOSi
MW: 203.3971 g/mol
MDL Number: MFCD18206129
CAS Number: 245660-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR