2-(3,5-Difluorophenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1414498-1G A1414498-250MG A1414498-5G
76660-626EA 26.53 USD
76660-626 76660-628 76660-630
2-(3,5-Difluorophenyl)-1,3,2-dioxaborinane 98%
2-(3,5-Difluorophenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₉H₉BF₂O₂
MW: 197.9744 g/mol
CAS Number: 684648-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR