3-(1,3,2-Dioxaborinan-2-yl)phenol 98%
Supplier: Ambeed
|
Warning
|
A1414183-1G
A1414183-250MG
76660-622EA
59.42
USD
76660-622
76660-624
3-(1,3,2-Dioxaborinan-2-yl)phenol 98%
3-(1,3,2-Dioxaborinan-2-yl)phenol
Precaución: for Research Use Only
Formula:
C₉H₁₁BO₃ MW: 177.9928 g/mol |
CAS Number:
2070921-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...