3-(Phenylethynyl)oxazolidin-2-one 98%

Supplier: Ambeed
Warning

A1414173-100MG A1414173-1G A1414173-250MG
76660-604EA 73.21 USD
76660-604 76660-606 76660-608
3-(Phenylethynyl)oxazolidin-2-one 98%
3-(Phenylethynyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.1947 g/mol
CAS Number: 888329-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR