2-(4-Methoxyphenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1411776-1G A1411776-5G A1411776-250MG
76660-596EA 54 USD
76660-596 76660-598 77874-775
2-(4-Methoxyphenyl)-1,3,2-dioxaborinane 98%
2-(4-Methoxyphenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
CAS Number: 155826-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR