(R)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride 97%

Supplier: Ambeed
Warning

A1411754-100MG A1411754-1G
76660-592EA 86 USD
76660-592 76660-594
(R)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride 97%
(R)-4-(Pyrrolidin-3-yl)morpholine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂O
MW: 229.1473 g/mol
MDL Number: MFCD18074308
CAS Number: 913702-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR