(Hexahydroindolizin-8a(1H)-yl)methanol 97%

Supplier: Ambeed
Warning

A1409945-100MG A1409945-250MG A1409945-50MG
76660-576EA 267 USD
76660-576 76660-578 76660-580
(Hexahydroindolizin-8a(1H)-yl)methanol 97%
(Hexahydroindolizin-8a(1H)-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₇NO
MDL Number: MFCD30003045
CAS Number: 1788874-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR