3-(tert-Butyl)-4-chlorobenzonitrile 97%

Supplier: Ambeed
Warning

A1409367-100MG A1409367-1G A1409367-250MG
76660-570EA 23 USD
76660-570 76660-572 76660-574
3-(tert-Butyl)-4-chlorobenzonitrile 97%
3-(tert-Butyl)-4-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN
MDL Number: MFCD29093068
CAS Number: 1369926-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR