2-((2R,3R)-3-(tert-Butyl)-4-(3,5-dicyclopentyl-2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)propan-2-ol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A1409277-100MG
A1409277-250MG
A1409277-25MG
A1409277-50MG
76660-556EA
291.75
USD
76660-556
76660-558
76660-560
76660-562
2-((2R,3R)-3-(tert-Butyl)-4-(3,5-dicyclopentyl-2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)propan-2-ol ≥98%, ee 99%
2-((2R,3R)-3-(tert-Butyl)-4-(3,5-dicyclopentyl-2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)propan-2-ol
Precaución: for Research Use Only
Formula:
C₃₂H₄₅O₄P MW: 524.6711 g/mol |
MDL Number:
MFCD32899518 CAS Number: 2416226-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...